missing translation for 'onlineSavingsMsg'
Learn More

Urea, 99%, (White crystalline powder or pellets/Ultra Pure), MP Biomedicals

Product Code. 11422002 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 lb.
Unit Size:
5lb
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422002 5 lb. 5lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11422002 Supplier MP Biomedicals Supplier No. 0210569505
  • ONLINE EXCLUSIVE PRICE
    108.00 €
    101.85 € / 5lb
    Save 6.15 €
    5% Off
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Color White
Assay Percent Range 99%
Quantity 5 lb.
Formula Weight 60.1
Grade Ultrapure
Physical Form Crystalline Powder or Pellets
Assay 4X Crystallized
Chemical Name or Material Urea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.