missing translation for 'onlineSavingsMsg'
Learn More

Urea, BP, USP, 99.0-100.5%, 99.0-101.0% (calc. dry substance), Honeywell Fluka™

Product Code. 16655743 Shop All Honeywell Fluka Products
Change view
Click to view available options
:
25 kg
Product Code.
16655743 25 kg
1 options
This item is not returnable. View return policy

Product Code. 16655743

Brand: Honeywell Fluka 15604H25KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 57-13-6
CAS Max % 1
Color White
Density 1.32 g/cm3 (20°C)
pH 9 to 10
Molecular Formula CH4N2O
Quantity 25 kg
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILES NC(N)=O
IUPAC Name urea
Molecular Weight (g/mol) 60.06
Grade Reagent Pharmacopeia Organics
Chemical Name or Material Urea
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.