missing translation for 'onlineSavingsMsg'
Learn More

Urea, Certified AR for Analysis, Fisher Chemical™

Product Code. 10578260 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
3kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10578260 3 kg Plastic powder jar 3kg
10102790 500 g Plastic powder jar 500g
10687422 5 kg Plastic keg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10578260 Supplier Fisher Chemical Supplier No. U/0500/63
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CH4N2O, CAS Number-57-13-6, carbamimidic acid, alphadrate, carbonyldiamide, isourea, urevert, ureaphil, pseudourea, ureophil, carbonyldiamine, carbamide, 5kg, CHEBI:48376, White, 60.06g/mol, XSQUKJJJFZCRTK-UHFFFAOYSA-N, 131 deg.C, 8022, 60.056, Plastic keg, 7.5 to 9.5, Solid, 1176, C(-O)(N)N

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number 8022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 131°C
Color White
pH 7.5 to 9.5
Packaging Plastic powder jar
Quantity 3 kg
Formula Weight 60.06g/mol
Vapor Pressure 1.25mmHg 25
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.