missing translation for 'onlineSavingsMsg'
Learn More

Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2

Product Code. 10420321 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10420321 250 g Glass bottle 250g
10266620 1 kg Glass bottle 1kg
11924381 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10420321 Supplier Thermo Scientific Acros Supplier No. 241022500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-43-6 | CH6N2O3 | 94.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-43-6 , 7722-88-5
Molecular Formula CH6N2O3
Molecular Weight (g/mol) 94.07
InChI Key AQLJVWUFPCUVLO-UHFFFAOYSA-N
Synonym urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid
PubChem CID 31294
ChEBI CHEBI:75178
IUPAC Name hydrogen peroxide;urea
SMILES C(=O)(N)N.OO

Specifications

Melting Point 75.0°C to 85.0°C
Color White
Packaging Glass bottle
Linear Formula CO(NH2)2·H2O2
Quantity 250 g
Fieser 16,379; 17,401
Merck Index 15,1784
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: alcohol and ether decomposes the product to h2o2, and urea, soluble in ethylene glycol
Stabilizer 1% sodium pyrophosphate
Formula Weight 94.07
Percent Purity ≥33.5% (H2O2)
Physical Form Tablets
Chemical Name or Material Urea hydrogen peroxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Urea hydrogen peroxide (1 g tablets), stabilized, contains 35 wt% H2O2
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.