missing translation for 'onlineSavingsMsg'
Learn More

Urea hydrogen peroxide adduct, 97%

Product Code. p-7046318
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11368835

Brand: Thermo Scientific Alfa Aesar L13940.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-43-6 | CH6N2O3 | 94.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Urea hydrogen peroxide is an antiseptic, disinfectant and bleaching agent used in pharmaceuticals and cosmetics respectively. It is used in the whitening of teeth and relieves minor inflammation of gums, oral mucosal surfaces and lips. It finds application in the preparation of plastics, blue print development and starch modification. In addition to this, it is used as a source of hydrogen peroxide easily handled in the laboratory.

Solubility
Soluble in water.

Notes
Incompatible with acetic anhydride, aniline and carboxylic acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-43-6
Molecular Formula CH6N2O3
Molecular Weight (g/mol) 94.07
MDL Number MFCD00013119
InChI Key AQLJVWUFPCUVLO-UHFFFAOYSA-N
Synonym urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid
PubChem CID 31294
ChEBI CHEBI:75178
IUPAC Name hydrogen peroxide;urea
SMILES C(=O)(N)N.OO

Specifications

Melting Point ∽90°C (decomposition)
Odor Odorless
Linear Formula H2NCONH2·H2O2
Quantity 25 g
UN Number UN1511
Beilstein 3680414
Sensitivity Very Hygroscopic
Merck Index 14,1782
Solubility Information Soluble in water.
Formula Weight 94.07
Percent Purity 97%
Chemical Name or Material Urea hydrogen peroxide adduct
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrogen peroxide--urea
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Urea, compound withhydrogen peroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.