missing translation for 'onlineSavingsMsg'
Learn More

Urea, MP Biomedicals™

Product Code. 11424682 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424682 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424682 Supplier MP Biomedicals Supplier No. 02191450.5
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in the solubilization and denaturation of proteins. Urea, MP Biomedicals is a principal protein metabolite end product of nitrogen metabolism in most mammals. It is formed by the enzymatic reactions of the Kreb's cycle and is the major product for th

Urea in solution is in equilibrium with ammonium cyanate. The form that reacts with protein amino groups is isocyanic acid. Urea in the presence of heat and protein leads to carbamylation of the proteins.

  • Carbamylation by isocyanic acid interferes with protein characterization because isocyanic acid reacts with the amino terminus of proteins, preventing N-terminal sequencing
  • Isocyanic acid also reacts with side chains of lysine and arginine residues, resulting in a protein that is unsuitable for many enzymatic digests
  • Carbamylation often leads to confusing results from peptides having unexpected retention times and masses

Urea is a mild agent usually used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. It is also useful for renaturing proteins from samples already denatured with 6M guanidine chloride, such as inclusion bodies.

  • It may be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form
  • It is commonly used in the extraction of the mitochondrial complex, in isoelectric focusing and two-dimensional electrophoresis, and in acetic acid-urea PAGE gels

Other applications include:

  • In cell or tissue culture media, to increase the osmolality
  • As fertilizer, because of the easy availability of nitrogen
  • In animal feeds
  • Reacted with aldehydes to make resins and plastics
  • Condensed with malonic ester to form barbituric acid
  • In the paper industry, to soften cellulose
  • As a diuretic
  • To enhance the action of sulfonamides
  • As an antiseptic

Specifications

CAS 57-13-6
Melting Point 132°C to 135°C
Density 1.335g/cm3 at 20°C(Lit.)
Boiling Point 135°C (literature) (decomposition)
Molecular Formula CH4N2O
Quantity 500 g
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
Solubility Information One gram dissolves in 1mL water, 10mL 95% ethanol (100mg/mL), 1mL boiling 95% ethanol, 20mL absolute ethanol (50mg/mL), 6mL methanol (166mg/mL), 2mL glycerol (500mg/mL); Soluble in concentrated hydrochloric acid; almost insoluble in chloroform, ether.
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILES C(=O)(N)N
IUPAC Name urea
Molecular Weight (g/mol) 60.056
PubChem CID 1176
ChEBI CHEBI:48376
Formula Weight 60.06
Vapor Pressure 0.000016hPa at 25°C (Lit.)
Percent Purity ≥99%
Grade ACS
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.