missing translation for 'onlineSavingsMsg'
Learn More

Urea, U.S.P., J.T. Baker™

Product Code. 15568274 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
500g
Product Code. Quantity Packaging unitSize
15568274 2.5 kg Poly Bottle 2.5kg
15558274 500 g Poly Bottle 500g
2 options
This item is not returnable. View return policy

Product Code. 15568274

Brand: Avantor J.T.Baker 420605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1L = 1.32kg
Formula Weight 60.06
Grade USP
Chemical Name or Material Urea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.