missing translation for 'onlineSavingsMsg'
Learn More

Urea, Ultra Pure, 5lb, MP Biomedicals

Product Code. 11471702 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 lb.
Unit Size:
5lb
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471702 5 lb. 5lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11471702 Supplier MP Biomedicals Supplier No. 04821527
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

57-13-6 Purity: ≥99% Heavy metals (as Pb): <<0.002 ppm O.D. (260nm) of a 7M solution: <<0.05 Protein denaturant

Specifications

CAS 57-13-6
Molecular Formula CH4N2O
Quantity 5 lb.
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILES C(=O)(N)N
IUPAC Name urea
Molecular Weight (g/mol) 60.056
PubChem CID 1176
ChEBI CHEBI:48376
Grade Ultrapure
Chemical Name or Material Urea
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.