missing translation for 'onlineSavingsMsg'
Learn More

Urethane, TRC

Product Code. 30457247
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457247 5 g 5g
30452781 25 g 25g
30450520 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457247 Supplier Toronto Research Chemicals Supplier No. TRCU8253005G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-79-6
Molecular Formula C3 H7 N O2
Molecular Weight (g/mol) 89.09
Synonym Ethyl aminoformate, Ethyl carbamate, Ethylurethane, Leucethane, NSC 746, O-Ethylurethane, Pracarbamine, Urethan, Urethane
IUPAC Name ethyl carbamate
SMILES CCOC(=O)N

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H7NO2/c1-2-6-3(4)5/h2H2,1H3,(H2,4,5)
Formula Weight 89.0477
Chemical Name or Material Ethyl carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.