missing translation for 'onlineSavingsMsg'
Learn More

Uric acid, 99% max., MP Biomedicals™

Product Code. 11487431 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487431 500 g 500g
11467431 10 g 10g
11477431 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487431 Supplier MP Biomedicals Supplier No. 0210321590
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 69-93-2
Molecular Formula C5H4N4O3
Molecular Weight (g/mol) 168.112
InChI Key LEHOTFFKMJEONL-UHFFFAOYSA-N
Synonym uric acid, urate, 2,6,8-trioxypurine, lithic acid, 2,6,8-trihydroxypurine, 2,6,8-trioxopurine, 1h-purine-2,6,8-triol, 8-hydroxyxanthine, 7,9-dihydro-1h-purine-2,6,8 3h-trione, 1h-purine-2,6,8 3h-trione, 7,9-dihydro
PubChem CID 1175
ChEBI CHEBI:17775
IUPAC Name 7,9-dihydro-3H-purine-2,6,8-trione
SMILES C12=C(NC(=O)N1)NC(=O)NC2=O

Specifications

Color White
Density 1.89g/mL
Assay Percent Range 99% max.
Quantity 500 g
Formula Weight 168.11
Physical Form Crystalline Powder
Chemical Name or Material Uric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.