missing translation for 'onlineSavingsMsg'
Learn More

Uric acid, +99%, Thermo Scientific Chemicals

Product Code. 10205560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10205560 25 g Plastic bottle 25g
10327400 100 g Plastic bottle 100g
10607134 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10205560 Supplier Thermo Scientific Acros Supplier No. 171290250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Uric acid, CAS # 69-93-2, is a common oxopurine metabolite, and a normal component of urine.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Uric acid is a purine-based metabolite found in humans, mice and E. coli
• Uric acid can accumulate in excess leading to diseases such as hyperuricemia
• Effects of excess uric acid accumulation include hypertension, arthritis, and renal issues
Applications

• Useful as a biochemical standard
• Suitable for use in the study of hyperuricemia and gout
• Uric acid may also play other roles in cell metabolism and physiology
• Intended for research use only

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-93-2
Molecular Formula C5H4N4O3
Molecular Weight (g/mol) 168.112
MDL Number MFCD00005712
InChI Key LEHOTFFKMJEONL-UHFFFAOYSA-N
Synonym uric acid, urate, 2,6,8-trioxypurine, lithic acid, 2,6,8-trihydroxypurine, 2,6,8-trioxopurine, 1h-purine-2,6,8-triol, 8-hydroxyxanthine, 7,9-dihydro-1h-purine-2,6,8 3h-trione, 1h-purine-2,6,8 3h-trione, 7,9-dihydro
PubChem CID 1175
ChEBI CHEBI:17775
IUPAC Name 7,9-dihydro-3H-purine-2,6,8-trione
SMILES C12=C(NC(=O)N1)NC(=O)NC2=O

Specifications

Color White
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Quantity 25 g
Formula Weight 168.11
Percent Purity ≥99.0% (ex N)
Physical Form Fine Powder
Chemical Name or Material Uric acid, 99+%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.