missing translation for 'onlineSavingsMsg'
Learn More

Uric Acid, TRC

Product Code. 30009661
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009661 10 g 10g
30007728 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009661 Supplier Toronto Research Chemicals Supplier No. TRCU82920010G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-93-2
Molecular Formula C5 H4 N4 O3
Molecular Weight (g/mol) 168.11
Synonym 7,9-Dihydro-1H-purine-2,6,8(3H)-trione (ACI), Uric acid (8CI), 1H-Purine-2,6,8-triol, 2,3,6,7,8,9-Hexahydro-1H-purine-2,6,8-trione, 2,6,8-Trihydroxypurine, 2,6,8-Trioxopurine, 2,6,8-Trioxypurine, Lithic acid, NSC 3975, Purine-2,6,8(1H,3H,9H)-trione, 1H-Purine-2,6,8(3H)-trione, 7,9-dihydro- (9CI, ACI)
IUPAC Name 7,9-dihydro-3H-purine-2,6,8-trione
SMILES O=C1NC(=O)C2=C(NC(=O)N2)N1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H4N4O3/c10-3-1-2(7-4(11)6-1)8-5(12)9-3/h(H4,6,7,8,9,10,11,12)
Formula Weight 168.0283
Chemical Name or Material Uric Acid Free Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.