missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Uridine 5'-monophosphate disodium salt

Product Code. 16489366
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489366 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16489366 Supplier MedChemExpress Supplier No. HYW013175
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Uridine 5'-monophosphate disodium salt is component used for RNA synthesis.

Specifications

Molecular Weight (g/mol) 368.14 g/mol
Product Type Natural Products
Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Shipping Condition Room temperature
Purity 98.0%
Quantity 100 mg
Solubility Information H2O : ≥ 50 mg/mL (135.82 mM)
CAS 3387-36-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.