missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Uridine-5'-monophosphate disodium salt, 99%

Product Code. 11498517
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498517 25 g 25g
11488517 5 g 5g
11408527 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498517 Supplier Thermo Scientific Chemicals Supplier No. A18601.14
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3387-36-8 | C9H11N2Na2O9P | 368.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3387-36-8
Molecular Formula C9H11N2Na2O9P
Molecular Weight (g/mol) 368.15
MDL Number MFCD00149422
InChI Key KURVIXMFFSNONZ-IJRYJGAHNA-L
Synonym uridine-5'-monophosphate disodium salt, uridine 5'-monophosphate, disodium salt, 5'-uridylicaciddisodiumsalt
PubChem CID 123134060
IUPAC Name [(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
SMILES [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O

Specifications

Melting Point ∼209°C (decomposition)
Quantity 25 g
Beilstein 3582885
Formula Weight 368.15
Percent Purity 99%
Chemical Name or Material Uridine-5'-monophosphate disodium salt

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.