missing translation for 'onlineSavingsMsg'
Learn More

Uridine 5’-Monophosphate Disodium Salt, TRC

Product Code. 30136673
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136673 10 g 10g
30136199 5 g 5g
17965075 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136673 Supplier Toronto Research Chemicals Supplier No. TRCU83000510G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3387-36-8
Molecular Formula C9 H11 N2 O9 P . 2 Na
Molecular Weight (g/mol) 368.14
Synonym Uridine 5'-Monophosphate Disodium, 5'-Uridylic acid, disodium salt (7CI,8CI,9CI), 5'-UMP disodium salt, Disodium 5'-UMP, Disodium 5'-uridylate, Disodium UMP, Disodium uridine 5'-monophosphate, Disodium uridine 5'-phosphate, Disodium uridine monophosphate, NSC 20257, UMP disodium salt, Uridine 5'-(dihydrogen phosphate) disodium salt, Uridine 5'-monophosphate disodium salt, Uridylic acid disodium salt, 5'-Uridylic acid, sodium salt (1:2)
IUPAC Name disodium;[(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])[O-])N2C=CC(=O)NC2=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H13N2O9P.2Na/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H,10,12,15)(H2,16,17,18);;/q;2*+1/p-2/t4-,6-,7-,8-;;/m1../s1
Formula Weight 367.9998
Chemical Name or Material Uridine-5'-monophosphate disodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.