missing translation for 'onlineSavingsMsg'
Learn More

Uridine 5'-Monophosphate, TRC

Product Code. 30138217
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138217 1 g 1g
30135308 5 g 5g
30005938 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138217 Supplier Toronto Research Chemicals Supplier No. TRCU8300201G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-97-9
Molecular Formula C9H13N2O9P
Molecular Weight (g/mol) 324.18
Synonym 5'-Uridylic Acid, Uridylic Acid, 5'-UMP, UMP, Uridine 5'-(Dihydrogen Phosphate), Uridine 5'-Phosphate, Uridine 5'-Phosphoric Acid, Uridine Monophosphate, Uridine Phosphate;
IUPAC Name ((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate
SMILES O[C@H]1[C@@H](O)[C@H](N(C=C2)C(NC2=O)=O)O[C@@H]1COP(O)(O)=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H13N2O9P/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18/h1-2,4,6-8,13-14H,3H2,(H,10,12,15)(H2,16,17,18)/t4-,6-,7-,8-/m1/s1
Formula Weight 324.04
Chemical Name or Material Uridine 5'-Monophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.