missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Uridine 5'-triphosphate, trisodium salt, hydrate, 90%

Product Code. 10793031 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10793031

Brand: Thermo Scientific Chemicals 226310010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19817-92-6 | C9H12N2Na3O15P3 | 550.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point >140.0°C
CAS 19817-92-6
Color White
pH 3.0 to 5.0 (0.5% aq. soln.)
Physical Form Powder
Quantity 1 g
Assay Percent Range 88% min. (HPLC)
Molecular Formula C9H12N2Na3O15P3
MDL Number MFCD11045990
Flash Point >110°C
Merck Index 10, 9688
Synonym uridine-5'-triphosphate trisodium salt dihydrate, uridine 5'-triphosphate trisodium salt dihydrate, trisodium uridine 5'-triphosphate dihydrate, 550.0866 anhydrous, c9h12n2na3o15p3.xh2o, uridine 5'-tetrahydrogen triphosphate , sodium salt, hydrate 1:3:2, trisodium dihydrate utp trianion, sodium 5'-o-hydroxyphosphinato oxy phosphinato oxy phosphinato uridine hydrate 3:1:2, trisodium, 2r,3s,4r,5r-5-2,4-dioxopyrimidin-1-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl hydrogen phosphate, dihydrate, uridine 5'-triphosphate, trisodium salt, hydrate
Solubility Information Solubility in water: 70%- 80% (20°C). Other solubilities: sparingly soluble in alcohol
InChI Key MMJGIWFJVDOPJF-ZKGVITBDNA-K
SMILES [Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)N1C=CC(=O)NC1=O
IUPAC Name trisodium;(2R,3R,4S,5R)-2-(2,4-dioxopyrimidin-1-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
Molecular Weight (g/mol) 550.09
PubChem CID 131675549
Formula Weight 550.1
Percent Purity 90%
Infrared Spectrum Authentic
Loss on Drying 8% max. (105°C, 5 hrs)
Packaging Glass bottle
Chemical Name or Material Uridine 5′-triphosphate, trisodium salt, hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.