missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Uridine

Product Code. 16438766
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438766 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438766 Supplier MedChemExpress Supplier No. HYB1449
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Uridine (β-Uridine) is a glycosylated pyrimidine-analog containing uracil attached to a ribose ring (or more specifically, aribofuranose) via a β-N1-glycosidic bond.

Specifications

Molecular Weight (g/mol) 244.2 g/mol
Product Type Natural Products
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 99.96%
Quantity 100 mg
Solubility Information H2O : ≥ 100 mg/mL (409.50 mM) ∣DMSO : 50 mg/mL (204.75 mM∣Need ultrasonic)
For Use With (Application) Metabolism-protein/nucleotide metabolism
CAS 58-96-8
Synonym β-Uridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.