missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Uridine, 99%

Product Code. 11402268
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11402268 100 g 100g
11482258 5 g 5g
11492258 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11402268 Supplier Thermo Scientific Chemicals Supplier No. A15227.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Uridine, 99%, C9H12N2O6, CAS Number-58-96-8, uracil, 1-beta-d-ribofuranosyl, uracil riboside, d-uridine, beta-uridine, 1-beta-d-ribofuranosyluracil, uridin, unii-whi7hq7h85, urd, b-uridine, uridine, 100g, 754904, 200-407-5, 244.2, DRTQHJPVMGBUCF-DHRBYNEYSA-N | CAS: 58-96-8 | C9H12N2O6 | 244.203 g/mol

Uridine plays a vital role in the glycolysis pathway of galactose. It is used as a precursor in the production of CDP-choline. It is an important nutrient and widely used as a dietary supplement. It improves brain cholinergic functions and hepatic mitochondrial function in certain liver toxins. It plays a major role in pain physiology and brain energy utilization to maintain ATP production under restricted oxygen conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uridine plays a vital role in the glycolysis pathway of galactose. It is used as a precursor in the production of CDP-choline. It is an important nutrient and widely used as a dietary supplement. It improves brain cholinergic functions and hepatic mitochondrial function in certain liver toxins. It plays a major role in pain physiology and brain energy utilization to maintain ATP production under restricted oxygen conditions.

Solubility
Soluble in water, dimethylsulfoxide, and methanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-96-8
Molecular Formula C9H12N2O6
Molecular Weight (g/mol) 244.203
MDL Number MFCD00006526
InChI Key DRTQHJPVMGBUCF-DHRBYNEYSA-N
Synonym uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
PubChem CID 89495556
IUPAC Name 1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O

Specifications

Melting Point 166°C to 169°C
Assay Percent Range 0.99
Quantity 100 g
Beilstein 754904
Merck Index 149877
Solubility Information Soluble in water,dimethylsulfoxide,and methanol.
Formula Weight 244.2
Percent Purity 99%
Chemical Name or Material Uridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.