missing translation for 'onlineSavingsMsg'
Learn More

Urocanic acid, MedChemExpress

Product Code. 16451189
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451189 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451189 Supplier Medchem Express Supplier No. HY113008
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Urocanic acid, produced in the upper layers of mammalian skin, is a major absorber of ultraviolet radiation (UVR).

Urocanic acid, produced in the upper layers of mammalian skin, is a major absorber of ultraviolet radiation (UVR).

Specifications

Chemical Name or Material Urocanic acid
CAS 104-98-3
Color White
Purity Grade Notes Research
Molecular Formula C6H6N2O2
Quantity 1 g
Solubility Information DMSO : 50 mg/mL (362.00 mM; Need ultrasonic) ∣H2O : 2 mg/mL (14.48 mM; Need ultrasonic)
SMILES O=C(O)/C=C/C1=CN=CN1
Molecular Weight (g/mol) 138.12
Formula Weight 138.12
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.