missing translation for 'onlineSavingsMsg'
Learn More

Urocanic Acid, TRC

Product Code. 30449562
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449562 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449562 Supplier Toronto Research Chemicals Supplier No. TRCU847063500MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Urocanic Acid
CAS 104-98-3
Molecular Formula C6 H6 N2 O2
Quantity 500 mg
Synonym 2-Propenoic acid','3-(1H-imidazol-4-yl)- (9CI), Imidazole-4-acrylic acid (8CI), 3-(1H-Imidazol-5-yl)-2-propenoic acid, 3-(1H-Imidazol-4-yl)-2-propenoic acid, 3-(1H-Imidazol-4-yl)acrylic acid, 3-(4-Imidazolyl)acrylic acid, 4-Imidazolylacrylic acid, 5-Imidazoleacrylic acid, NSC 66357, Urocanic acid, Urocaninic acid
InChI Formula InChI=1S/C6H6N2O2/c9-6(10)2-1-5-3-7-4-8-5/h1-4H,(H,7,8)(H,9,10)
SMILES OC(=O)C=Cc1c[nH]cn1
IUPAC Name 3-(1H-imidazol-4-yl)prop-2-enoic acid
Molecular Weight (g/mol) 138.12
Formula Weight 138.0429
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.