missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Urolithin A
GREENER_CHOICE

Product Code. 18756986
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18756986 10 mg 10mg
18709556 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18756986 Supplier Tocris Bioscience™ Supplier No. 6762/10
  • ONLINE EXCLUSIVE PRICE
    121.00 €
    114.45 € / 10mg
    Save 6.55 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

PI 3-K/Akt/mTOR signaling pathway inhibitor; blocks phosphorylation of Akt and p70S6K in pancreatic ductal adenocarcinoma (PDAC)

Urolithin A is a PI 3-K/Akt/mTOR signaling pathway inhibitor; blocks phosphorylation of Akt and p70S6K in pancreatic ductal adenocarcinoma (PDAC). Inhibits proliferation and migration and promotes apoptosis of PDAC cell lines in vitro (IC50 values are 10.23 - 62.28 μM). Suppresses tumor growth and improves survival in mice bearing pancreatic tumor xenografts. Also blocks amyloid-β fibrillation in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,8-Dihydroxy-6H-dibenzo[b,d]pyran-6-one
CAS 1143-70-0
Quantity 10 mg
Molecular Formula C13H8O4
Formula Weight 228.2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.