missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ursodeoxycholic acid, 99%

Product Code. 10431251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10431251 5 g Glass bottle 5g
10052660 1 g Glass bottle 1g
10259273 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10431251 Supplier Thermo Scientific Chemicals Supplier No. 208590050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 128-13-2 | C24H40O4 | 392.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-13-2
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003680
InChI Key RUDATBOHQWOJDD-UZVSRGJWSA-N
Synonym ursodeoxycholic acid, ursodiol, actigall, ursofalk, ursolvan, delursan, ursodeoxycholate, urso, urso forte, ursodesoxycholic acid
PubChem CID 31401
ChEBI CHEBI:9907
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])[C@@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Melting Point 200°C to 206°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 10074
Specific Rotation Condition +60.00 (20.00°C c=2,C2H5OH)
Specific Rotation +58° to +62° (20°C, 589nm) (c=2, ethanol)
IUPAC Name (4R)-4-[(1R,3aS,3bR,4S,5aS,7R,9aS,9bS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Formula Weight 392.57
Percent Purity ≥98.5% (Acid)
Physical Form Solution
Chemical Name or Material Ursodeoxycholic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.