missing translation for 'onlineSavingsMsg'
Learn More

Ursodeoxycholic Acid, 99%, MP Biomedicals™

Product Code. 11459721 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459721 25 g 25g
11449721 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459721 Supplier MP Biomedicals Supplier No. 0215825225
  • ONLINE EXCLUSIVE PRICE
    558.00 €
    529.20 € / 25g
    Save 28.80 €
    5% Off
 Restricted Item
Estimated Shipment: 26-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 128-13-2
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003680
InChI Key RUDATBOHQWOJDD-UZVSRGJWSA-N
Synonym ursodeoxycholic acid, ursodiol, actigall, ursofalk, ursolvan, delursan, ursodeoxycholate, urso, urso forte, ursodesoxycholic acid
PubChem CID 31401
ChEBI CHEBI:9907
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])[C@@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Assay Percent Range 99%
Quantity 25 g
IUPAC Name (4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Formula Weight 392.6
Physical Form Solid
Chemical Name or Material Ursodeoxycholic Acid
compliance-icons
Product Identifier
  • Ursodeoxycholic Acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ursodeoxycholic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.