Learn More
SI Analytics™ PC-Keyboard
Product Code. 11398300 Shop All SI Analytics Products
Change view
Click to view available options
For Use With (Equipment):
TTITRONIC and TitroLine
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
11398300 TTITRONIC and TitroLine Each
1 options
Product Code. 11398300 Supplier SI Analytics™ Supplier No. 285220410
  • ONLINE EXCLUSIVE PRICE
    167.00 €
    107.00 € / Pack of 1
    Save 60.00 €
    35% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SI Analytics™ PC-Keyboard includes a USB plug to allow it to be connected to TitroLine™ and TITRONIC™ instruments.
  • With USB plug connection
  • Suitable for use with TitroLine™ 5000/6000, TitroLine™ 7000 , TitroLine™ 7500 KF, TitroLine™ 7500 KF trace, TitroLine™ 7750 and TITRONIC™ 300/500
TRUSTED_SUSTAINABILITY
Specifications
Connector Types USB
Type Keyboard
For Use With (Equipment) TTITRONIC and TitroLine