missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Service Fittings for Laboratory Enclosures: These service fittings allow water, gas, compressed air and electrical or data connections to be integrated directly inside Erlab enclosures, so that operators have all the utilities they need at the point of use, without compromising containment.
- Safe Integration Inside the Working Volume:
- Components designed for installation in Erlab enclosure panels
- Sealed, leak-tight assembly that preserves the containment of the working volume
- Corrosion-resistant materials suitable for laboratory environments
- Positioning studied to keep the working surface clear and ergonomic
- Electrical accessories compliant with applicable safety requirements
- Practical, Ready-to-Install Solutions: Supplied as a complete kit with the required sealing and fixing components, these fittings can be installed on new equipment or retrofitted to an existing enclosure according to Erlab instructions.
- Key Benefits:
- Utilities available directly at the point of use.
- Leak-tight installation that preserves enclosure containment.
- Corrosion-resistant, easy-to-clean materials.
- Ready-to-install kit designed for Erlab equipment.
- Improved operator comfort and workstation efficiency.
Specifications
Specifications
| Product Type | USB Port |
| For Use With (Equipment) | Solis |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?