missing translation for 'onlineSavingsMsg'
Learn More

Erlab USB Port inside Right Side

Product Code. 31793975 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
Solis
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31793975 Solis Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31793975 Supplier Erlab Supplier No. NORT000424
  • ONLINE EXCLUSIVE PRICE
    484.00 €
    451.00 € / Pack of 1
    Save 33.00 €
    6% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An integrated solution to simplify the use of peripheral devices due to the USB Port conveniently located on our BSC.

  • Service Fittings for Laboratory Enclosures: These service fittings allow water, gas, compressed air and electrical or data connections to be integrated directly inside Erlab enclosures, so that operators have all the utilities they need at the point of use, without compromising containment.
  • Safe Integration Inside the Working Volume:
    • Components designed for installation in Erlab enclosure panels
    • Sealed, leak-tight assembly that preserves the containment of the working volume
    • Corrosion-resistant materials suitable for laboratory environments
    • Positioning studied to keep the working surface clear and ergonomic
    • Electrical accessories compliant with applicable safety requirements
  • Practical, Ready-to-Install Solutions: Supplied as a complete kit with the required sealing and fixing components, these fittings can be installed on new equipment or retrofitted to an existing enclosure according to Erlab instructions.
  • Key Benefits:
    • Utilities available directly at the point of use.
    • Leak-tight installation that preserves enclosure containment.
    • Corrosion-resistant, easy-to-clean materials.
    • Ready-to-install kit designed for Erlab equipment.
    • Improved operator comfort and workstation efficiency.

Specifications

Product Type USB Port
For Use With (Equipment) Solis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.