missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ USB Power Supply, PIPETMAN™ M

Product Code. 15555516 Shop All Gilson Products
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15555516 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15555516 Supplier Gilson™ Supplier No. F807015
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This USB Power Supply is an accessory for PIPETMAN™ M and PIPETMAN™ M Connected electronic pipettes.

  • USB power supply for PIPETMAN™ M and PIPETMAN™ M Connected electronic pipettes
TRUSTED_SUSTAINABILITY

Specifications

Material Plastic
Sterility Non-sterile
Color Black
Type Power Supply
Disposable Reusable
For Use With (Equipment) PIPETMAN™ M and PIPETMAN™ M Connected
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.