Learn More
SI Analytics™ USB Thermo Printer
Product Code. 15352367 Shop All SI Analytics Products
Change view
Click to view available options
Length (Metric):
112 mm
Unit Size:
Each
Product Code. Length (Metric) Unit Size
15352367 112 mm Each
1 options
Product Code. 15352367 Supplier SI Analytics™ Supplier No. 285220480
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SI Analytics™ USB Thermo Printer is an accessory for use with Titronic™ 500 and TitroLine™ 7000/7500/7750/7800.
  • For use with TitroLine™ 6000/7000/7500 KF/7500 KF trace/7750 and TITRONIC™ 500
TRUSTED_SUSTAINABILITY
Specifications
Connector Types USB
Type Thermo Printer
Length (Metric) 112 mm
Size 112 mm