Learn More
Memmert™ Usb User-Id Stick (With User-Id License), For Ico150Med
Product Code. 16326827 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
Units With TwinDISPLAY
Unit Size:
Each
Product Code. 16326827
Brand: Memmert™ ICO150medB33170
Only null left
Add to basket
Add to basket
Includes
User-ID License
Memmert™ USB User-ID Stick prevents undesired manipulation by unauthorized third parties.
  • USB User-ID stick (with User-ID License)
  • Oven-linked authorisation License (User-ID-programme) on Memory-stick
  • Prevents undesired manipulation by unauthorised third parties
  • When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
Description Only for units with TwinDISPLAY
For Use With (Equipment) Units With TwinDISPLAY
Type USB Flash Drive