missing translation for 'onlineSavingsMsg'
Learn More

Vilber Lourmat™ UV Cabinet without Lamp

Product Code. 11786628
Change view
Click to view available options
Quantity:
1Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11786628 1Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11786628 Supplier Vilber Lourmat™ Supplier No. 411000601
  • ONLINE EXCLUSIVE PRICE
    492.00 €
    191.76 € / Pack of 1
    Save 300.24 €
    61% Off
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rich, robust UV cabinet features light curtain, central sighting scope with anti-reflection UV protector, and 2 receptacles for 6 W lamp.

  • Portable UV Room 12 W
  • For 1 CCM plate 20 x 20cm
  • 2 x 6 W UV tubes
  • Light curtain, central sighting scope with anti-reflection UV protector and 2 receptacles for 6 W lamp

Specifications

Product Type UV Cabinet
Quantity 1Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.