Description
A 2mm flow cell for UV-LED monitor U9-L. Included with ÅKTA pure equipped with U9-L. Can be used as second UV monitor.
Specifications
Specifications
| For Use With (Equipment) | U9-L |
| Includes | ÄKTA pure equipped with U9-L. Can be used as second UV monitor |
| Quantity | 1 Each |