Learn More
ERLAB UV Decontamination
Product Code. 30484797 Shop All Erlab Products
Change view
Click to view available options
Voltage:
0/1 V
Unit Size:
Each
Product Code. Voltage Unit Size
30484797 0/1 V Each
1 options
Product Code. 30484797 Supplier ERLAB Supplier No. NORT000475
  • ONLINE EXCLUSIVE PRICE
    313.00 €
    290.00 € / Pack of 1
    Save 23.00 €
    7% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Installed in microbiological safety cabinets, provides an efficient method for sterilizing the workspace
  • UV Decontamination uses germicidal UV-C light to destroy microorganisms on surfaces within the cabinet
  • Automatically or manually operated for timed decontamination cycles between work sessions
  • Promotes aseptic conditions for sensitive microbiological or cell culture applications
  • Reduces the need for harsh chemical disinfectants inside the workspace
  • Integrated safety sensors disable UV light when the sash is open, protecting users from exposure
  • Recommended for daily sterilization routines in biosafety environments
  • UV lamp is easy to replace and provides consistent performance over time
Specifications
Voltage 0/1 V