missing translation for 'onlineSavingsMsg'
Learn More

Vilber Lourmat™ UV Lamp

Product Code. 11776518
Click to view available options
Lighting Intensity:
1100 mW/cm2
1350/1100 mW/cm2
1780 mW/cm2
2300 mW/cm2
265/350 mW/cm2
340 mW/cm2
400 mW/cm2
700 mW/cm2
710 mW/cm2
Wattage:
4 W
6 W
8 W
15 W
30 W
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11776518

Brand: Vilber Lourmat™ 311100451

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Single wavelength UV lamp with filter that is an ideal solution for research and enumeration of E. coli.

  • The dimensions of the lamps are fixed according to the power
  • 6W Lamp: L = 27.6cm x D = 6cm x H = 8.5cm
  • Double wavelength UV lamp with filter
  • Lamp 2 x 4 W

Specifications

Wattage 8 W
Wavelength 365/254 nm
Lighting Intensity 265/350 mW/cm2
Packaging Quantity 1 Ea.
Product Type UV Lamp
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.