missing translation for 'onlineSavingsMsg'
Learn More

Vilber Lourmat™ UV Lamp

Product Code. 11830430
Change view
Click to view available options
Lighting Intensity:
1100 mW/cm2
1350/1100 mW/cm2
1780 mW/cm2
2300 mW/cm2
265/350 mW/cm2
340 mW/cm2
400 mW/cm2
700 mW/cm2
710 mW/cm2
Wattage:
4 W
6 W
8 W
15 W
30 W
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lighting Intensity Wattage Unit Size
11830430 710 mW/cm2 6 W Each
11776518 265/350 mW/cm2 8 W Each
11820430 340 mW/cm2 4 W Each
11860380 400 mW/cm2 4 W Each
11870380 700 mW/cm2 6 W Each
11800250 1100 mW/cm2 15 W Each
11796658 1350/1100 mW/cm2 30 W Each
11766668 1780 mW/cm2 30 W Each
11746668 2300 mW/cm2 30 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11830430 Supplier Vilber Lourmat™ Supplier No. 311100611
  • ONLINE EXCLUSIVE PRICE
    594.00 €
    457.55 € / Pack of 1
    Save 136.45 €
    22% Off
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Single wavelength UV lamp with filter that is an ideal solution for research and enumeration of E. coli.

  • Single wavelength UV lamp with filter
  • 6 W UV lamp
  • Rapid visualization of CCM plates requiring UV revelation
  • Power supply
  • Complete with UV tube and filter
  • Research and enumeration of E. coli
  • UV lamp 254nm

Specifications

Voltage 230 V
Wattage 6 W
Wavelength 254 nm
Depth (Metric) 85 mm
Electrical Requirements 6 W, 230 V, 50 Hz
Height (Metric) 60 mm
Hertz 50 Hz
Length (Metric) 276 mm
Lighting Intensity 710 mW/cm2
Packaging Quantity 1 Ea.
Product Type UV Lamp
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.