Learn More
Vilber Lourmat™ UV Lamp Stand
Product Code. 11726678
Change view
Click to view available options
For Use With (Equipment):
15w UV lamp
4/8w Handheld UV Lamp
4/8w UV lamp
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11726678 15w UV lamp Each
11840240 4/8w UV lamp Each
11850240 4/8w Handheld UV Lamp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11726678 Supplier Vilber Lourmat™ Supplier No. 913020041
  • ONLINE EXCLUSIVE PRICE
    229.00 €
    151.39 € / Pack of 1
    Save 77.61 €
    33% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Rich and robust table stand for ultraviolet lamps 4/8 W.
  • Stand for UV lamps 4/8W
  • Double wavelength lamp
  • 254/365 nm Intensity 
  • Lamp 2 x 6 W
  • M, 2 table stand
Specifications
For Use With (Equipment) 15w UV lamp