Learn More
Vilber Lourmat™ UV Lamp Stand
Click to view available options
For Use With (Equipment):
15w UV lamp
4/8w Handheld UV Lamp
4/8w UV lamp
Unit Size:
Each
Description
- Stand for UV lamps 4/8W
- Double wavelength lamp
- 254/365 nm Intensity
- Lamp 2 x 6 W
- M, 2 table stand
Specifications
Specifications
| Type | UV Lamp Stand |
| Description | Stand for UV lamps 4/8W |
| For Use With (Equipment) | 4/8w UV lamp |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido