Learn More
Syngene™ UV Safety Shield
Product Code. 16524351 Shop All Syngene Products
Change view
Click to view available options
Size:
Small
Large
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16524351 Large Each
16534351 Small Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16524351 Supplier Syngene™ Supplier No. UVSHIELDLARGE
  • ONLINE EXCLUSIVE PRICE
    234.00 €
    211.98 € / Pack of 1
    Save 22.02 €
    9% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Safety shield designed to offer UV protection.
  • The wearing of safety equipment according to laboratory regulations is recommended
  • Large Shield Dimensions: 14 W x 24 in. H
Specifications
Product Line Syngene
For Use With (Application) UV Protection
Product Type Safety Shield
For Use With (Equipment) GBox Chemi XX6 and XX9
Includes UV Safety Shield, 24 in. high
Size Large
Voltage 110/230 V