Learn More
Eppendorf™ UVette™ Starter Set
Product Code. 10081992 Shop All Eppendorf Products
Change view
Click to view available options
Type:
UVette™ Starter Set
Unit Size:
1 set
Product Code. Type Unit Size
10081992 UVette™ Starter Set 1 set
1 options
Product Code. 10081992 Supplier Eppendorf™ Supplier No. 4099100007
  • ONLINE EXCLUSIVE PRICE
    110.55 €
    102.00 € / 1 sets
    Save 8.55 €
    7% Off
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Individually packaged UVettes™ are DNA, RNA and protein free, and are protected from scratches and dust
Individually packaged UVettes™ are DNA, RNA and protein free, and are protected from scratches and dust.
  • Disposable cuvette for UV light measurements
  • Fully transparent plastic for all VIS applications
  • UV and VIS transparent between 220nm to 1, 600nm
  • Suitable for measuring small volumes (≥50μL)
  • Individually blister-packed for contamination free work processes
  • Volume markings for 500μL and 1,000μL
  • Optimal filling guaranteed by funnel shaped tube base
  • Frosted grip allows for labelling of the cuvette
  • Deep-lying optical window prevents scratches
  • Samples with a normal concentration can be measured using the 10mm optical path length, for samples with a higher concentration, simply rotate the UVette™ by 90°C and use the shorter 2mm optical path length
  • Can be used as is with the BioPhotometer, and with all other spectrophotometers with the aid of an adapter
  • Adapter aperture: 1.5mm x 1.5mm (h x w)
TRUSTED_SUSTAINABILITY
Specifications
Description Starter Set
Type UVette™ Starter Set
For Use With (Application) For measurements in the UV/Vis range with transparency from 220 nm to 1600 nm