Learn More
Uvex™ Astrospec 2.0 Replacement Shield
Product Code. 15385892 Shop All Uvex Products
Change view
Click to view available options
:
Product Type: Replacement lens
Product Code.
15385892 Product Type: Replacement lens
1 options
Product Code. 15385892 Supplier Uvex™ Supplier No. 9168055
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Replacement lens for Astrospec 2.0
  • Polycarbonate lens, clear
  • UV 400, 2-1.2 W 1 F DIN CE
  • Coating uvex supravision sapphire
  • TRUSTED_SUSTAINABILITY
    Specifications
    Product Type Replacement lens
    Lens Material Polycarbonate