Learn More
uvex C300 Foam Cut Protection Gloves
Product Code. 15317092 Shop All Uvex Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15317092 10 Each
15337092 7 Each
15347092 8 Each
15357092 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15317092 Supplier uvex Supplier No. 6054410
  • ONLINE EXCLUSIVE PRICE
    17.65 €
    16.46 € / Pack of 1
    Save 1.19 €
    6% Off
In Stock
Add to basket
Edge
Add to basket
Uvex C300 Foam gloves set standards for comfort, tactility and efficiency while also providing level 3/C cut protection. High-performance elastomer coating on the palm and fingertips enhances grip capabilities in dry or slightly damp applications.
  • Gloves provide level 3/C blade cut resistance, abrasion resistance, and high tear resistance
  • High-performance elastomer (HPE) coating on the palm and fingers for enhanced grip capabilities
  • Soft-grip foam coating provides mechanical abrasion resistance
  • Highly flexible with very good tactile sensation
  • Uvex Climazone technology reduces excessive moisture and warmth
  • 3D Ergo technology for a secure fit
  • Silicone-free according to the imprint test
TRUSTED_SUSTAINABILITY
Specifications
Coated Area Palm and fingertips
Coating Material HPE
Color Gray
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Latex Free Yes
Material HPPE, Polyamide, Glass, Bamboo Viscose
Product Type Cut-resistant Gloves
Size 10
EN388 Standard 3X42C
For Use With (Application) Automotive industry, assembly, maintenance, metalworking, shipping/logistics, sorting, glass handling, and sheet metal processing
Glove Cuff Style Knit
Series C300 Foam