Learn More
Uvex™ Pheos Eyewear
Product Code. 15396072 Shop All Uvex Products
Change view
Click to view available options
Frame Color:
Black, gray
Cool Gray
Green, Gray
Yellow, Black
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frame Color Unit Size
15396072 Black, gray Each
15366072 Cool Gray Each
15376072 Green, Gray Each
15326082 Yellow, Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15396072 Supplier Uvex™ Supplier No. 9192280
  • ONLINE EXCLUSIVE PRICE
    13.45 €
    12.59 € / Pack of 1
    Save 0.86 €
    6% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
More than just a common spectacle - the modern fashion-look makes it a welcome companion at everyday work
  • Perfect all-round protection, large field of vision through duo spherical lens, outstanding ventilation thanks to integrated uvex climazone temple concept
  • Coating uvex supravision excellence
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance EN 170
    Lens Type Uvex™ supravision excellence
    Lens Coating Anti-fog, Scratch-resistant
    Description Shield-lens spectacle
    Lens Tint Clear
    Product Type Ergonomic Safety Glasses
    Frame Color Black, gray
    Material Polycarbonate
    Disposable Reusable
    Lens Material Polycarbonate
    Frame Material Plastic
    UV Protection UV400
    Frame Type Standard