missing translation for 'onlineSavingsMsg'
Learn More

uvex Profapren CF33 Chemical Protecion Gloves

Product Code. 15316922 Shop All Uvex Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15316922

Brand: uvex 6011904

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

High-quality unsupported chloroprene safety gloves for protection against a broad spectrum of chemicals

  • Silicone-free safety gloves with excellent properties for protection against chemical and mechanical risks
  • Good combination of flexibility and strength
  • Resistant to many chemicals and solvents
  • EN 388:2016 (3131X)
  • EN ISO 374-1: 2016 / Type A (AKLMNO)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 388:2016 (3 1 3 1 X), 374-1:2016 /Type A (A K L M N O)
Coating Material Polychloroprene
Color Blue
Latex Free Yes
Material Cotton
Product Type Chemical-resistant Gloves
Size 10
Thickness (Metric) 0.75 mm
For Use With (Application) Chemical industry, painting/coating, metal work (cleaning)
Model CF33
Palm Type Napped
Length (Metric) 33 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.