Learn More
uvex Safety Glasses Sports Style
Product Code. 12364729 Shop All Uvex Products
Change view
Click to view available options
Product Type:
Sports-style Spectacles
Unit Size:
Each
Product Code. Product Type unitSize
12364729 Sports-style Spectacles Each
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12364729 Supplier uvex Supplier No. 9160520
Stock Up Your Lab
Use promo code 28007
View all promotions
In Stock
Add to basket
Edge
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Features adjustable temple length
Innovative styling and design, with adjustable temple length and lens inclination.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint Transparent
Product Type Sports-style Spectacles
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard