missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Unilite Thermo Safety Gloves

Product Code. p-7160283 Shop All Uvex Products
Change view
Click to view available options
Size:
8
9
10
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15387132 10 Each
15317142 8 Each
15327142 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15387132 Supplier Uvex™ Supplier No. 6059310
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Robust hand protection for cold working conditions

  • Despite its extremely robust coating, the uvex unilite thermo stays warm and flexible even in extreme cold
  • The two-layer material composition of acrylic and new wool ensures a pleasant hand climate and good wearer comfort at all times
  • Polymer coating on palm and fingertips that is flexible at low temperatures
  • Good grip in dry and slightly damp areas
  • Very good thermal insulation in direct contact with cold objects
  • Good tactile sensation, good fit, highly flexible, and good wearer comfort
  • Certified to EN 388:2016 (3 1 3 1 X) and EN 511 (0 1 0)
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance 388:2016 (3 1 3 1 X), 511 (0 1 0), 388 (3 2 3 1)
    Coated Area Palm and fingertips
    Coating Material Polymer
    Color Black
    Exterior Finish Smooth
    Hand Ambidextrous
    Material Elastane
    Product Type Cold-resistant Gloves
    Size 10
    For Use With (Application) Work at low temperatures, refrigerated warehouses, and forklift truck driving
    Glove Cuff Style Knit
    Liner Material Acrylic/ new wool
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.