missing translation for 'onlineSavingsMsg'
Learn More

Uvinul A Plus-d4, TRC

Product Code. 30457136
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457136 10 mg 10mg
30454270 5 mg 5mg
30453214 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30457136 Supplier Toronto Research Chemicals Supplier No. TRCU85030110MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethylaminohydroxybenzoyl hexyl benzoate D4 (phenyl-2,3,4,5-D4)
Molecular Formula C24 D4 H27 N O4
Quantity 10 mg
Synonym Diethylaminohydroxybenzoyl Hexyl Benzoate D4 (phenyl 2,3,4,5-D4)
InChI Formula InChI=1S/C24H31NO4/c1-4-7-8-11-16-29-24(28)20-13-10-9-12-19(20)23(27)21-15-14-18(17-22(21)26)25(5-2)6-3/h9-10,12-15,17,26H,4-8,11,16H2,1-3H3/i9D,10D,12D,13D
SMILES [2H]c1c([2H])c([2H])c(C(=O)c2ccc(cc2O)N(CC)CC)c(C(=O)OCCCCCC)c1[2H]
IUPAC Name hexyl 2,3,4,5-tetradeuterio-6-[4-(diethylamino)-2-hydroxybenzoyl]benzoate
Molecular Weight (g/mol) 401.5319
Formula Weight 401.2504
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.