missing translation for 'onlineSavingsMsg'
Learn More

UVP Clear UV Blocking Covers

Product Code. 12317553
Change view
Click to view available options
For Use With (Equipment):
3uv™ transilluminators
Benchtop transilluminators
Standard Format UV Transilluminators
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
12317553 Standard Format UV Transilluminators Each
11852172 3uv™ transilluminators Each
12823908 Benchtop transilluminators Each
3 options
This item is not returnable. View return policy

Product Code. 12317553

Brand: UVP 19011201

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

For Use With (Application) Gel electrophoresis
Color Clear
For Use With (Equipment) Standard Format UV Transilluminators
Includes Uv Blocking Cover
Quantity each
Size 18 x 13 in.
Product Type UV Blocking Cover
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.