Learn More
Sperian™ V-Maxx Acetate Safety Goggle
Product Code. 11806980
Change view
Click to view available options
Lens Tint:
Clear
Unit Size:
Each
Product Code. Lens Tint Unit Size
11806980 Clear Each
1 options
Product Code. 11806980 Supplier Sperian™ Supplier No. 1007506
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Certifications/Compliance ISO 9001 / 2000
Lens Coating Anti-fog
Lens Tint Clear
Product Type Safety Goggles
For Use With (Application) Painting, General Use and Protection Against Fogging
Headband Material Neoprene
No. per Pack 1 Ea.
Ventilation Details Indirect
Material Acetate