missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ V-MOVE-90 Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas with electronic open/close doors.
4349.71 € - 6061.61 €
Specifications
Certifications/Compliance | EN 14470-1, TRGS 510 |
---|---|
Product Type | Safety storage cabinet |
Door Style | Auto-close |
For Use With (Application) | Storage of flammable hazardous substances |
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
15939541
|
Asecos™
3503500135103 |
5 x Shelves: 50 kg each, Inserts: 50 kg each, Sumps: 13 L each, Maximum load: 240 kg |
6061.61 €
Each |
Estimated Shipment: 01-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916371
|
Asecos™
3508504035100 |
4 x Shelves: 50 kg each, Insert: 50 kg, Sump: 13 L, Maximum load: 120 kg |
4349.71 €
Each |
Estimated Shipment: 01-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15826501
|
Asecos™
3508604135104 |
4 x Shelves: 50 kg each, Inserts: 50 kg each, Sumps: 13 L each, Maximum load: 240 kg |
6020.80 €
Each |
Estimated Shipment: 01-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Optimum integration and use of space – with a depth of 86 cm, V-LINE cabinets now use the space next to laboratory fume cupboards and work benches that conventional hazardous material cabinets have left unused
- Fire resistance of 90 minutes
- Maximum comfort – vertical drawer technology allows access and visibility to all stored containers from both sides
- At the push of a button the vertical drawers open and close automatically
- Drawers glide softly and quietly out of and into the cabinet body without additional physical efforts
- The stop function of the vertical drawers detects resistances and eliminates the risk of being trapped
- No unauthorized use – doors lockable with cylinder lock and cabinets can be integrated by the customer into an existing master key system
- Locking status indicator (red/green) provided
- Always closed – serial automatic door closing after approx. 60 seconds with acoustic/visual signal before closing
- Possible interior equipment includes height-adjustable shelves, bottom collecting sumps and guard rail systems
- In accordance with EN 14470-1 and TRGS 510
- Robust construction and longevity – scratch-resistant outer body with powder-coated surface
- Ventilation – integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
EN 14470-1, TRGS 510 | |
Safety storage cabinet | |
Auto-close | |
Storage of flammable hazardous substances |