missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ V1 Series Visor
Description
- Steel mesh visor with a matt finish for extremely harsh working environments
- Optimum design ensures excellent strength and stability
- A visor for extremely harsh working environments
- Designed to fit all 3M™ Safety Helmets
- All mesh visors are tested and approved under the European Standard EN 371 and meet the requirements for increased robustness (S marking)
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | 3M™ Safety Helmets |
| Finish | Matte |
| Material | Steel Mesh |
| Product Type | Visor |
| Size (Metric) | 1.8 x 1.8 mm mesh size |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?